Skip to Content
Merck
All Photos(1)

Key Documents

860640P

Avanti

Sphingosine (d17:1)

Avanti Research - A Croda Brand 860640P, powder

Synonym(s):

D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H35NO2
CAS Number:
Molecular Weight:
285.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860640P-10mg)
pkg of 1 × 5 mg (860640P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860640P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCC

InChI

1S/C17H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17(20)16(18)15-19/h13-14,16-17,19-20H,2-12,15,18H2,1H3/b14-13+/t16-,17+/m0/s1

InChI key

RBEJCQPPFCKTRZ-LHMZYYNSSA-N

General description

Sphingosine (d17:1) or D-erythro-sphingosine is a 17-carbon sphingolipid, found in human skin. It has one carbon less than naturally occurring C18-sphingosine.

Application

Sphingosine (d17:1) has been used as a lipid standard in the analysis of sphingolipids by liquid chromatography-mass spectrometry (LC-MS) and tandem mass spectrometry.

Biochem/physiol Actions

Sphingosine acts a precursor for sphingosine-1-phosphate (S1P). Sphingosine C-17 is used as an internal standard in the analysis of sphingolipids by spectrometric methods.

Packaging

5 mL Amber Glass Screw Cap Vial (860640P-10mg)
5 mL Amber Glass Screw Cap Vial (860640P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A selective sphingosine kinase 1 inhibitor integrates multiple molecular therapeutic targets in human leukemia
Paugh SW, et al.
Blood, 112(4), 1382-1391 (2008)
Cytochrome c oxidase deficiency accelerates mitochondrial apoptosis by activating ceramide synthase 6
Schull S, et al.
Cell Death & Disease, 6(3), e1691-e1691 (2015)
Free sphingosines of human skin include 6-hydroxysphingosine and unusually long-chain dihydrosphingosines
Stewart ME and Downing DT
The Journal of Investigative Dermatology, 105(4), 613-618 (1995)
Evaluation of bioactive sphingolipids in 4-HPR-resistant leukemia cells
Apraiz A, et al.
BMC Cancer, 11(1), 477-477 (2011)
Guang-Yi Dai et al.
The Journal of biological chemistry, 295(3), 717-728 (2019-12-11)
Cellular membranes contain many lipids, some of which, such as sphingolipids, have important structural and signaling functions. The common sphingolipid glucosylceramide (GlcCer) is present in plants, fungi, and animals. As a major plant sphingolipid, GlcCer is involved in the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service