Skip to Content
Merck
All Photos(1)

Key Documents

810185P

Avanti

TopFluor PI(4)P

Avanti Research - A Croda Brand 810185P, powder

Synonym(s):

1-oleoyl-2-{6-[4-(dipyrrometheneboron difluoride)butanoyl]amino}hexanoyl-sn-glycero-3-phosphoinositol-4-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H88BF2N5O17P2
CAS Number:
Molecular Weight:
1142.01
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (with stopper and crimp cap (810185P-50ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810185P

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Phosphatidylinositol 4-phosphate (PI4P) is a precursor of phosphatidylinositol (4,5)-bisphosphate [PI(4,5)P2]. It is present in the membranes of Golgi apparatus.

Application

TopFluor PI(4)P has been used as fluorescently labeled phosphoinositide to examine phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase (PTEN) interaction with anionic lipids.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810185P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PI4P and PI (4, 5) P2 are essential but independent lipid determinants of membrane identity
Hammond GRV, et al.
Science, 337(6095), 727-730 (2012)
Xiaojun Shi et al.
Langmuir : the ACS journal of surfaces and colloids, 32(7), 1732-1741 (2016-02-02)
Phosphatidylinositol phosphate (PIP) lipids are critical to many cell signaling pathways, in part by acting as molecular beacons that recruit peripheral membrane proteins to specific locations within the plasma membrane. Understanding the biophysics of PIP-protein interactions is critical to developing
Biophysical methods for the characterization of PTEN/lipid bilayer interactions
Harishchandra RK, et al.
Methods, 77, 125-135 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service