610010
Avanti
PC Membranes 1.0um
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder PC Membrane, Polycarbonate Membranes 1.0μm 19mm (100/pk)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
General description
Polycarbonate Membrane for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Application
PC Membranes 1.0um has been used in the mini-extruder to prepare lipid vesicles for fluorescence spectroscopy.
Biochem/physiol Actions
Polycarbonate membranes are useful to prepare uniform and specific sized liposomes by ordered extrusion.
Components
Whatman® Nuclepore Hydrophilic Membrane 800319
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva
also commonly purchased with this product
Product No.
Description
Pricing
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Quantitative Fluorometric Approach for Measuring the Interaction of RhoGDI with Membranes and Rho GTPases
Johnson J, et al.
Methods in Molecular Biology, 107-119 (2012)
Preparation of liposomes of defined size distribution by extrusion through polycarbonate membranes
Olson F, et al.
Biochimica et Biophysica Acta - Biomembranes, 557(1), 9-23 (1979)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service