Z742517
Block for digital dry bath and MyBlock™
Holds 24 x 1.5 mL or 2.0 x tubes
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NB.77
Recommended Products
feature
Holds 24 x 1.5 mL or 2.0 x tubes
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSW1520
Related Categories
General description
For use with Benchmark Digital Dry Bath
- Material: high grade aluminum
- Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.