Z742324
2MAG MIX 4 MS multiple 4 point stirrer
AC/DC input 100-240 V, US 3-pin plug
Synonym(s):
induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer
About This Item
Recommended Products
material
stainless steel housing
stainless steel plate
feature
positions 4
28 W (4 x 7 W
4 step)
packaging
pack of 1 ea
manufacturer/tradename
2mag 30404
parameter
-10-50 °C temp. limit (at 80 percent humidity)
100-2000 rpm speed
W × D × H
245 mm × 425 mm × 38 mm
stirrer distance
140 mm
stirrer volume
1-3000 mL , per stirring position
weight
~8.9 kg
Looking for similar products? Visit Product Comparison Guide
General description
Powerful 4 stirring positions, 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept,
power reserves up to 3,000 ml per stirring point, individual
and independent speed setting for each stirring drive
(Master-Slave-Technology), also ON/OFF function,
synchronous speed mode for all positions, extremely
wide speed range of 100 to 2,000 rpm, jerk-free stirring
even at low speeds.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service