Key Documents
Z741033
Sigma-Aldrich
CJ Tee Syringe Adapter - CGA 180 Inlet
About This Item
UNSPSC Code:
12142200
NACRES:
NA.12
Pricing and availability is not currently available.
Quality Level
Related Categories
Application
Use to extract samples from gas cylinders. Please see attached bulletin for detailed instructions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)