Z560057
Levgo® smartSpatula® disposable PP laboratory spatula, Look for the V at the neck™
L 310 mm, green, macro
Synonym(s):
disposable spatula, macrospatula, spatula
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Recommended Products
material
polypropylene
packaging
pack of 150 ea
case of 3600 ea
manufacturer/tradename
LevGo Inc 17241
L
310 mm
color
green
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
General description
Versatile Double-ended polypropylene tool that can scoop, scrape, spoon, stir, collect, transfer, handle or manipulate a variety of chemicals and biological materials.
LevGo spatulas are widely used in commercial, university, diagnostic and pharmaceutical labs worldwide as well as by compounding pharmacists to produce customized medications.
LevGo spatulas are widely used in commercial, university, diagnostic and pharmaceutical labs worldwide as well as by compounding pharmacists to produce customized medications.
Features and Benefits
- The V design at the neck of the disposable lab spatula is a trademark of LevGo, Inc.
- No Searching, washing or contamination.
- 100% Polypropylene is autoclavable and resistant to dilute acids and bases.
- Polypropylene will not shatter after exposure to liquid nitrogen frozen samples.
- Hollow core uses less plastic than other spatulas and minimizes heat or cold transfer.
- SGS-Tested for US and EU food contact standards.
- Sustainability – LevGo plants a tree for every case sold.
Legal Information
US Patent 8,709,362
LevGo is a registered trademark of LevGo Inc.
Look for the V at the neck is a trademark of LevGo Inc.
smartSpatula is a registered trademark of LevGo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service