Key Documents
Z531480
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 600 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
availability
not available in EU
Overall H
600 mm
General description
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)