Key Documents
Z514918
Kimax® high strength centrifuge tube replacement cap
Synonym(s):
73802-24400
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
manufacturer/tradename
Kimble®
Features and Benefits
• Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving• PTFE faced/general purpose white rubber liners are highly resistant to chemical effects• Excellent for general laboratory use• Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.