Z263486
Wheaton straight-mouth bottles with cap, clear
clear soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
Synonym(s):
Wheaton bottles, bottles clear, clear bottles
About This Item
Recommended Products
material
(PTFE faced LDPE liner)
clear soda-lime glass bottle
round bottle
white polypropylene cap
Joint
Joints threaded neck (89-400)
sterility
non-sterile
feature
graduations: no
with cap
packaging
case of 12 ea
manufacturer/tradename
WHEATON® W216912
capacity
1000 mL
diam. × H
95 mm × 170 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Wide mouth design is great for large solid samples
- Clear, USP Type III soda-lime glass
- Straight walls allow for complete removal of contents
- Ideal for soil sampling and environmental applications
- Available with caps attached or bulk packs without caps
- HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification
- Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service