material
borosilicate glass
feature
graduations: no
volume
100 mL
Related Categories
General description
One upper and one lower hose connection on opposite sides. Without pouring spout. A 3/8 in. I.D. hose is specified. A vacuum is not used with this product. Hot or cold liquids are circulated through the jacket to control temperature within the beaker.
Outer Jacket O.D. 57mm x H 120mm.
Inner Vessel I.D. ~38mm x D 110mm.
Inner Vessel I.D. ~38mm x D 110mm.
Other Notes
Browse all glass beakers
Choose from one of the most recent versions:
Certificates of Analysis (COA)