Skip to Content
Merck
All Photos(1)

Key Documents

W412101

Sigma-Aldrich

Geranic acid

85%, stabilised

Synonym(s):

3,7-Dimethyl-2,6-octadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCO2H
CAS Number:
Molecular Weight:
168.23
FEMA Number:
4121
Beilstein:
1763804
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.081
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

description

Natural occurrence: tea, tomato, wine.

Assay

85%

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.484 (lit.)

bp

250 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble

density

0.97 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; floral; woody

SMILES string

C\C(C)=C\CC\C(C)=C\C(O)=O

InChI

1S/C10H16O2/c1-8(2)5-4-6-9(3)7-10(11)12/h5,7H,4,6H2,1-3H3,(H,11,12)/b9-7+

InChI key

ZHYZQXUYZJNEHD-VQHVLOKHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geranic acid is a monoterpenemainly used in flavor, fragrance, cosmetics and agro-industries. In vitro studies show that geranic acid exhibits strong inhibitory activity against Fusarium graminearum and Colletotrichum graminicola, the two main fungal pathogens of maize. It may be responsible for the potent tyrosinase inhibitory activity of lemongrass (Cymbopogon citratus).

Application

Possible uses: orange, tea, mint, ripe fruit and melon notes.

Biochem/physiol Actions

Odor (neat): sweet, floral green, geranium leaf-like, woody with a citrus orange body nuance.
Taste at 20 ppm: sweet, woody, leafy, fruity body with some mouthfeel effects.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo production of the monoterpenoid geranic acid by metabolically engineered Pseudomonas putida.
Mi J, et al.
Microbial cell factories, 13(1), 170-170 (2014)
Identification of geranic acid, a tyrosinase inhibitor in lemongrass (Cymbopogon citratus).
Masuda T, et al.
Journal of Agricultural and Food Chemistry, 56(2), 597-601 (2007)
T Yamaguchi et al.
Journal of the National Cancer Institute, 73(4), 903-907 (1984-10-01)
The antitumor effects of combinations of synthetic isoprenoids-decaprenylamine.HCl, N-(p-methylbenzyl)decaprenylamine.HCl [N-(PMB)-decaprenylamine.HCl], and decaprenoic acid-with anticancer agents were studied in male ICR mice with ascites sarcoma 180 (S180) and in male BALB/c mice with fibrosarcoma Meth A. Decaprenylamine.HCl, N-(PMB)-decaprenylamine.HCl, and decaprenoic acid
Mosciano, Gerard
Perfumer & Flavorist, 33, 66-66 (2008)
Ting Yang et al.
Metabolic engineering, 13(4), 414-425 (2011-02-08)
Many terpenoids are known to have antifungal properties and overexpression of these compounds in crops is a potential tool in disease control. In this study, 15 different mono- and sesquiterpenoids were tested in vitro against two major pathogenic fungi of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service