Skip to Content
Merck
All Photos(1)

Key Documents

W319902

Sigma-Aldrich

5-Methyl-2-phenyl-2-hexenal

≥96%, FG

Synonym(s):

cocoa hexenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH=C(C6H5)CHO
CAS Number:
Molecular Weight:
188.27
FEMA Number:
3199
EC Number:
Council of Europe no.:
10365
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.099
NACRES:
NA.21
Organoleptic:
bitter; chocolate; cocoa
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

vapor density

>1 (vs air)

Assay

≥96%

refractive index

n20/D 1.534 (lit.)

density

0.973 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

bitter; chocolate; cocoa

SMILES string

[H]C(C(C1=CC=CC=C1)=CCC(C)C)=O

InChI

1S/C13H16O/c1-11(2)8-9-13(10-14)12-6-4-3-5-7-12/h3-7,9-11H,8H2,1-2H3

InChI key

YURDCJXYOLERLO-UHFFFAOYSA-N

Application

  • Characterization of aroma active volatile components in roasted mullet roe.: This study identifies 5-Methyl-2-phenyl-2-hexenal as a key component contributing to the distinctive aroma profile of roasted mullet roe. This research provides insights into flavor science and the role of volatile compounds in food aroma, helping enhance product development in the food industry (Fang M et al., 2022).

  • A 90-day subchronic toxicity study of 5-methyl-2-phenyl-2-hexenal in F344 rats.: This toxicological evaluation assesses the safety of 5-Methyl-2-phenyl-2-hexenal, providing essential data for its use in various consumer products. Understanding its toxicological profile helps ensure safety in its application, crucial for regulatory approval and consumer safety (Ogawa K et al., 2021).

  • 5-Methyl-2-phenyl-2-hexenal in the scent gland secretion of male cave crickets, Troglophilus cavicola and T. neglectus (Ensifera: Rhaphidophoridae).: Describes the presence of 5-Methyl-2-phenyl-2-hexenal in the scent glands of cave crickets, suggesting its role in chemical communication and possibly in mating behavior. Such studies enhance our understanding of animal behavior and pheromone-based communication systems (Leis HJ et al., 1998).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alekandra Bojke et al.
Journal of microbiological methods, 169, 105835-105835 (2020-01-10)
In this study, a procedure was developed to determine aldehydes using headspace solid-phase microextraction (HS-SPME) followed by gas chromatography (GC) coupled with mass spectrometry (MS). The aldehydes selected for research had previously been identified in various species of insects. Minimal
Rowe, D.
Perfumer & Flavorist, 30, 20-20 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service