Skip to Content
Merck
All Photos(1)

Documents

W293415

Sigma-Aldrich

Propyl butyrate

natural, ≥95%, FG

Synonym(s):

n-Propyl-n-butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH2CH2CH3
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2934
Beilstein:
1745552
EC Number:
Council of Europe no.:
266c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.040
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥95%

refractive index

n20/D 1.4 (lit.)

bp

142-143 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; ethereal; sweet

SMILES string

CCCOC(=O)CCC

InChI

1S/C7H14O2/c1-3-5-7(8)9-6-4-2/h3-6H2,1-2H3

InChI key

HUAZGNHGCJGYNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural occurrence: Apple, banana, kiwi, melon, papaya, passion fruit, rum and strawberry.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service