Skip to Content
Merck
All Photos(1)

Key Documents

W245100

Sigma-Aldrich

Ethyl palmitate

≥95%, FG

Synonym(s):

Ethyl hexadecanoate, Palmitic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOC2H5
CAS Number:
Molecular Weight:
284.48
FEMA Number:
2451
Beilstein:
1782663
EC Number:
Council of Europe no.:
634
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.193
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.440 (lit.)

bp

192-193 °C/10 mmHg (lit.)

mp

24-26 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

creamy; milk; waxy

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC

InChI

1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3

InChI key

XIRNKXNNONJFQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl palmitate is a one of the major volatile compounds found in cooked rice and Luzhou-flavor raw liquor. It occurs naturally in licorice root and chamomile.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic synthesis of ethyl palmitate in supercritical carbon dioxide
Industrial & Engineering Chemistry Research, 43(7), 1568-1573 (2004)
Discrimination of different kinds of Luzhou-flavor raw liquors based on their volatile features
Zheng J, et al
Food Research International, 56, 77-84 (2014)
Kosei Eguchi et al.
Cell metabolism, 15(4), 518-533 (2012-04-03)
Consumption of foods high in saturated fatty acids (FAs) as well as elevated levels of circulating free FAs are known to be associated with T2D. Though previous studies showed inflammation is crucially involved in the development of insulin resistance, how
M A Diczfalusy et al.
European journal of biochemistry, 259(1-2), 404-411 (1999-01-23)
Fatty acid ethyl esters have been detected in high concentrations in organs commonly damaged by alcohol abuse and are regarded as being important non-oxidative metabolites of ethanol. The formation of fatty acid ethyl esters (FAEEs) has been ascribed to two
Ashanty M Pina-Rodriguez et al.
Journal of agricultural and food chemistry, 57(11), 4657-4662 (2009-05-06)
Amaranth oil is rich in linoleic, oleic, and palmitic acids. Structured lipids (SLs) with specific functional and nutritional characteristics can be prepared through chemical or enzymatic interesterification. The aim of this study was to increase the palmitic acid content at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service