TEM-HC200-CU-UL
Ultra-Thin Holey Carbon Film Supported Copper Grids
size 200 mesh, box of 25
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Recommended Products
packaging
box of 25
size
200 mesh
Related Categories
General description
A 4-6 nm ultra-thin holey carbon film supported grids. Copper grids, 200 Mesh, 25/box
Choose from one of the most recent versions:
Certificates of Analysis (COA)