Skip to Content
Merck
All Photos(1)

Key Documents

T27405

Sigma-Aldrich

Thianaphthene

95%

Synonym(s):

Benzo[b]thiophene, Benzothiophene, Thionaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6S
CAS Number:
Molecular Weight:
134.20
Beilstein:
80580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

bp

221-222 °C (lit.)

mp

30-33 °C (lit.)

density

1.149 g/mL at 25 °C (lit.)

SMILES string

c1ccc2sccc2c1

InChI

1S/C8H6S/c1-2-4-8-7(3-1)5-6-9-8/h1-6H

InChI key

FCEHBMOGCRZNNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Tyteca et al.
Journal of chromatography. A, 1358, 145-154 (2014-07-21)
Linear gradient programs are very frequently used in reversed phase liquid chromatography to enhance the selectivity compared to isocratic separations. Multi-linear gradient programs on the other hand are only scarcely used, despite their intrinsically larger separation power. Because the gradient-conformity
Susana O Ribeiro et al.
Materials (Basel, Switzerland), 11(7) (2018-07-14)
Different methodologies were used to increase the oxidative desulfurization efficiency of the Keggin phosphotungstate [PW12O40]3- (PW12). One possibility was to replace the acid proton by three different ionic liquid cations, forming the novel hybrid polyoxometalates: [BMIM]₃PW12 (BMIM as 1-butyl-3-methylimidazolium), [BPy]₃PW12
Adrian G Clark et al.
Journal of chromatography. A, 1421, 154-161 (2015-04-25)
An improved multiple flame photometric detector (mFPD) is introduced, based upon interconnecting fluidic channels within a planar stainless steel (SS) plate. Relative to the previous quartz tube mFPD prototype, the SS mFPD provides a 50% reduction in background emission levels
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of
Alan P Graves et al.
Journal of medicinal chemistry, 48(11), 3714-3728 (2005-05-27)
Molecular docking is widely used to predict novel lead compounds for drug discovery. Success depends on the quality of the docking scoring function, among other factors. An imperfect scoring function can mislead by predicting incorrect ligand geometries or by selecting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service