SYNC184464C
Synthware™ chromatography column with PTFE stopcock and coarse fritted disc
top joint: ST/NS 24/40, I.D. × L 40.0 mm × 457 mm
Synonym(s):
Synthware C184464C, glass chromatograpy column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Joint
joint: ST/NS top24/40
grade
for chromatography
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C184464C
I.D. × L
40.0 mm × 457 mm
Looking for similar products? Visit Product Comparison Guide
General description
General purpose column has a PTFE stopcock and a top outer joint. With a coarse porosity fritted disc to support column packing. The section below disc is carefully minimized to prevent re-mixing of components already separated. All Synthware columns are heavy wall designed to increase safety.
This product includes coarse fritted disc.
This product includes coarse fritted disc.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service