Skip to Content
Merck
All Photos(1)

Key Documents

QBD10866

Sigma-Aldrich

SPDP-dPEG®36-acid

>95% (HPLC)

Synonym(s):

OPSS-PEG1500-acid, OPSS-PEG36-acid, SPDP-PEG-acid, SPDP-PEG36-COOH, SPDP-PEG36-acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C83H158N2O39S2
Molecular Weight:
1872.26
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

S(Sc1ncccc1)CCC(=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)O

InChI

1S/C83H158N2O39S2/c86-81(5-80-125-126-82-3-1-2-6-85-82)84-7-9-90-11-13-92-15-17-94-19-21-96-23-25-98-27-29-100-31-33-102-35-37-104-39-41-106-43-45-108-47-49-110-51-53-112-55-57-114-59-61-116-63-65-118-67-69-120-71-73-122-75-77-124-79-78-123-76-74-121-72-7

InChI key

YETQZNCXLDFCHR-UHFFFAOYSA-N

Features and Benefits

SPDP-dPEG36-acid is a crosslinking reagent designed to conjugate free amines and sulfhydryls. The single molecular weight (i.e., monodispersed) PEG crossbridge between the two functional groups is 115 atoms (137.8 Å) long. Using a suitable carbodiimide, such as EDC, the propionic acid group reacts with free primary amines to form amide bonds. The pyridyl disulfide (SPDP, also known as OPSS) functional group reacts with free thiols to form disulfide bonds. This product is useful for crosslinking peptides, proteins, and small molecules, and modifying surfaces with suitable reactive sites. SPDP-dPEG36-acid is a monodisperse equivalent of the polymeric PEG1500.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service