Skip to Content
Merck
All Photos(1)

Key Documents

QBD10784

Sigma-Aldrich

Biotin-dPEG®11-azide

Synonym(s):

Polyethylene glycol, alpha-(2-azidoethyl)-omega-[2-[[5-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]ethoxy]-poly(oxy-1,2-ethanediyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H64N6O13S
CAS Number:
Molecular Weight:
796.97
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1NC(C2N1)CSC2CCCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-])=O

InChI

1S/C34H64N6O13S/c35-40-37-6-8-44-10-12-46-14-16-48-18-20-50-22-24-52-26-28-53-27-25-51-23-21-49-19-17-47-15-13-45-11-9-43-7-5-36-32(41)4-2-1-3-31-33-30(29-54-31)38-34(42)39-33/h30-31,33H,1-29H2,(H,36,41)(H2,38,39,42)

InChI key

WETRQYMCASPBIY-UHFFFAOYSA-N

Features and Benefits

Biotin-dPEG®11-azide is a single molecular weight dPEG® that permits biotin labeling with precise spacing to molecules containing an alkyne using well-known, highly popular click chemistry. By inserting an alkyne function into the target molecule, site specific labeling can occur using either copper(I) catalyzed or strain-catalyzed click chemistry. The single molecular weight nature of the dPEG® spacer (50.4 Å) allows the user to precisely tailor the distance between the biotin and the target molecule in order to receive optimal binding between biotin and avidin.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service