All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C30H48N2O17
CAS Number:
Molecular Weight:
708.71
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
>90%
form
solid or viscous liquid
reaction suitability
reagent type: cross-linking reagent
reactivity: amine reactive
polymer architecture
shape: linear
functionality: homobifunctional
shipped in
ambient
storage temp.
−20°C
SMILES string
O=C(ON1C(CCC1=O)=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON2C(CCC2=O)=O)=O
Features and Benefits
Bis-dPEG®9-NHS ester is a homobifunctional dPEG® crosslinking product which allows conjugation to two different moieties, each containing free amines. The single molecular weight dPEG® spacer (35.7 Å) provide precise distance control between conjugated moieties.
Legal Information
Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
11 - Combustible Solids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service