Skip to Content
Merck
All Photos(1)

Key Documents

P63603

Sigma-Aldrich

2,5-Pyridinedicarboxylic acid

98%

Synonym(s):

Isocinchomeronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO4
CAS Number:
Molecular Weight:
167.12
Beilstein:
131697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

242-247 °C (dec.) (lit.)

SMILES string

OC(=O)c1ccc(nc1)C(O)=O

InChI

1S/C7H5NO4/c9-6(10)4-1-2-5(7(11)12)8-3-4/h1-3H,(H,9,10)(H,11,12)

InChI key

LVPMIMZXDYBCDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Pyridinedicarboxylic acid can be used:
  • In the synthesis of diorganotin(IV) 2,5-pyridinedicarboxylates.
  • In the preparation of three-dimensional coordination polymers of lanthanide 2,5-pyridinedicarboxylates by hydrothermal method.
  • As an organocatalyst in the synthesis of 1,5-benzodiazepine derivatives via one pot three-component reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noemí Contreras-Pereda et al.
Ultrasonics sonochemistry, 70, 105292-105292 (2020-08-05)
Large blue rectangular crystals of the 2D layered coordination polymer 1 have been obtained. The interest for this complex is two-fold. First, complex 1 is made of 2D layers packing along the (0-11) direction favored by the presence of lattice
Syntheses, structures, and photoluminescence of three-dimensional lanthanide coordination polymers with 2, 5-pyridinedicarboxylic acid.
Huang Y, et al.
Journal of Solid State Chemistry, 181(8), 1731-1737 (2008)
2, 6-Pyridinedicarboxylic acid as organocatalyst for the synthesis of 1, 5-benzodiazepines through one-pot reaction.
Lal M, et al.
Tetrahedron Letters, 54(32), 4264-4272 (2013)
Self-Assembly of Diorganotin (IV) Oxides (R= Me, n Bu, Ph) and 2, 5-Pyridinedicarboxylic Acid to Polymeric and Trinuclear Macrocyclic Hybrids with Porous Solid-State Structures: Influence of Substituents and Solvent on the Supramolecular Structure.
Garcia-Zarracino R and Hopfl H
Journal of the American Chemical Society, 127(9), 3120-3130 (2005)
Debal Kanti Singha et al.
Dalton transactions (Cambridge, England : 2003), 46(34), 11344-11354 (2017-08-16)
Herein, a mixed metal coordination polymer, {(H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service