Skip to Content
Merck
All Photos(2)

Documents

P20955

Sigma-Aldrich

2-Phenylbutyric acid

98%

Synonym(s):

(±)-2-Phenylbutyric acid, α-Ethylphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(C6H5)COOH
CAS Number:
Molecular Weight:
164.20
Beilstein:
509876
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

270-272 °C (lit.)

mp

39-42 °C (lit.)

SMILES string

CCC(C(O)=O)c1ccccc1

InChI

1S/C10H12O2/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,11,12)

InChI key

OFJWFSNDPCAWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin A Cho et al.
PloS one, 9(11), e110086-e110086 (2014-11-08)
Dystonia is a neurological disorder in which sustained muscle contractions induce twisting and repetitive movements or abnormal posturing. DYT1 early-onset primary dystonia is the most common form of hereditary dystonia and is caused by deletion of a glutamic acid residue
Burkhard Lehner et al.
BMC cancer, 14, 495-495 (2014-07-10)
Growing evidence exists that the neoplastic stromal cell population (GCTSC) within giant cell tumors (GCT) originates from mesenchymal stem cells (MSC). In a previous study we identified a microRNA signature that differentiates between these cell types. Five differentially expressed microRNAs
Dima Albals et al.
Electrophoresis, 35(19), 2807-2818 (2014-07-02)
A generic chiral separation strategy for the analysis of acidic compounds in CEC is proposed in completion of an earlier defined strategy for nonacidic compounds. The screening step of this strategy uses a 45 mM ammonium formate (pH 2.9)/ACN (35/65

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service