N17807
5-Nitroisatin
97%
Synonym(s):
5-Nitroindole-2,3-dione, 5-Nitrosotin, NSC 525798
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H4N2O4
CAS Number:
Molecular Weight:
192.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
251 °C (dec.) (lit.)
SMILES string
[O-][N+](=O)c1ccc2NC(=O)C(=O)c2c1
InChI
1S/C8H4N2O4/c11-7-5-3-4(10(13)14)1-2-6(5)9-8(7)12/h1-3H,(H,9,11,12)
InChI key
UNMYHYODJHKLOC-UHFFFAOYSA-N
Related Categories
Application
Reactant for preparation of:
- Potential antibacterial and antifungal agents
- spiro[indole-thiazolidinones] as a medicinally important scaffold
- Potential antimycobacterial agents
- Anticonvulsant agents
- Inhibitors of human transglutaminase 2
- Anthelmintic agents
- Anti-human immunodeficiency virus (HIV) agents
- Antimicrobial agents
- Potential antitubercular agents
- Inhibitors of acetylcholinesterase
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hideki Iwamoto et al.
Molecular therapy oncolytics, 2, 15020-15020 (2015-01-01)
"Angiogenic switch off" is one of the ideal therapeutic concepts in the treatment of cancer. However, the specific molecules which can induce "angiogenic switch off" in tumor have not been identified yet. In this study, we focused on von Hippel-Lindau
Chaitany Jayprakash Raorane et al.
Biomolecules, 10(8) (2020-08-23)
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service