All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
EC Number:
MDL number:
UNSPSC Code:
12141910
PubChem Substance ID:
Recommended Products
description
gray
Assay
≥99.999%
form
pellets
resistivity
1.2 μΩ-cm, 0°C
bp
684.9 °C (lit.)
mp
217 °C (lit.)
density
4.81 g/mL at 25 °C (lit.)
SMILES string
[Se]
InChI
1S/Se
InChI key
BUGBHKTXTAQXES-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kai-Oliver Feldmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(27), 9697-9712 (2015-05-12)
Although a fairly large number of binary group 15/16 element cations have been reported, no example involving phosphorus in combination with a group 16 element has been synthesized and characterized to date. In this contribution is reported the synthesis and structural characterization
Claire M Weekley et al.
Nutrients, 5(5), 1734-1756 (2013-05-24)
Determining the speciation of selenium in vivo is crucial to understanding the biological activity of this essential element, which is a popular dietary supplement due to its anti-cancer properties. Hyphenated techniques that combine separation and detection methods are traditionally and
Joseph L Jacobson et al.
Environmental health perspectives, 123(8), 827-833 (2015-03-11)
Although prenatal methylmercury exposure has been linked to poorer intellectual function in several studies, data from two major prospective, longitudinal studies yielded contradictory results. Associations with cognitive deficits were reported in a Faroe Islands cohort, but few were found in
Mark Denham et al.
Stem cells (Dayton, Ohio), 33(6), 1759-1770 (2015-03-11)
The caudal neural plate is a distinct region of the embryo that gives rise to major progenitor lineages of the developing central and peripheral nervous system, including neural crest and floor plate cells. We show that dual inhibition of the
Matthew Loxham et al.
Toxicological sciences : an official journal of the Society of Toxicology, 145(1), 98-107 (2015-02-13)
We have previously shown that underground railway particulate matter (PM) is rich in iron and other transition metals across coarse (PM10-2.5), fine (PM2.5), and quasi-ultrafine (PM0.18) fractions and is able to generate reactive oxygen species (ROS). However, there is little
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service