Skip to Content
Merck
All Photos(2)

Key Documents

GF89918500

Bismuth

foil, 25x25mm, thickness 0.25mm, 100%

Synonym(s):

Bismuth, BI000220

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

100%

form

foil

manufacturer/tradename

Goodfellow 899-185-00

resistivity

129 μΩ-cm, 20°C

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bismuth in medicine.
Hongzhe Sun et al.
Metal ions in biological systems, 41, 333-378 (2004-06-23)
Ruiguang Ge et al.
Metallomics : integrated biometal science, 4(3), 239-243 (2012-02-24)
Helicobacter pylori causes various gastric diseases, such as gastritis, peptic ulcerations and gastric cancer. Bismuth-based triple or quadruple therapies have been commonly recommended for the treatment of H. pylori infections. Up to now, the molecular mechanisms by which bismuth inhibits
Syntheses of biodegradable and biocompatible polymers by means of bismuth catalysts.
Hans R Kricheldorf
Chemical reviews, 109(11), 5579-5594 (2009-09-26)
Hong Lu et al.
European journal of gastroenterology & hepatology, 25(10), 1134-1140 (2013-06-20)
Antimicrobial resistance has continued to undermine many popular anti-Helicobacter pylori therapies. Antibiotic resistance to commonly used anti-H. pylori drugs in China has increased markedly, making China an ideal site to identify regimens that remain effective despite widespread antimicrobial resistance. Bismuth
Jorge A R Salvador et al.
Future medicinal chemistry, 4(11), 1495-1523 (2012-08-04)
In recent years, the chemical potential of bismuth and bismuth compounds has been actively exploited. Bismuth salts are known for their low toxicity, making them potential valuable reagents for large-scale synthesis, which becomes more obvious when dealing with products such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service