Skip to Content
Merck
All Photos(2)

Key Documents

GF74207347

Indium

wire reel, 1m, diameter 1.5mm, as drawn, 99.999%

Synonym(s):

Indium, IN005140

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

99.999%

form

wire

manufacturer/tradename

Goodfellow 742-073-47

resistivity

8.37 μΩ-cm

L × diam.

1 m × 1.5 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The significance of chromosomal aberrations in indium-111-labeled lymphocytes.
M L Thakur et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 25(8), 922-927 (1984-08-01)
[Scintigraphy with indium 111-labeled blood platelets].
C Kessler et al.
Deutsche medizinische Wochenschrift (1946), 109(48), 1853-1859 (1984-11-30)
Mikio Nakajima et al.
Congenital anomalies, 48(4), 145-150 (2008-11-06)
Indium, a precious metal classified in group 13 (IIIB) in the periodic table, has been used increasingly in the semiconductor industry. Because indium is a rare metal, technology for indium recycling from transparent conducting films for liquid crystal displays is
Indium-111 imaging in osteomyelitis and neuroarthropathy. Review and case report.
L M Techner et al.
Journal of the American Podiatric Medical Association, 76(1), 23-29 (1986-01-01)
[Use of 113m Indium as marker of compounds in the scintigraphy of the brain, lungs, liver, spleen, kidneys, placenta and heart].
A Mikolajków et al.
Polski tygodnik lekarski (Warsaw, Poland : 1960), 28(31), 1200-1201 (1973-07-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service