G10601
Glyoxylic acid monohydrate
98%
Synonym(s):
Formylformic acid, Oxoethanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
HCOCO2H · H2O
CAS Number:
Molecular Weight:
92.05
Beilstein:
969535
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
bp
100 °C (lit.)
mp
49-52 °C (lit.)
SMILES string
[H]O[H].[H]C(=O)C(O)=O
InChI
1S/C2H2O3.H2O/c3-1-2(4)5;/h1H,(H,4,5);1H2
InChI key
MOOYVEVEDVVKGD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Employed in the synthesis of a sulfinylmaleate, which serves as an efficient dienophile for enantioselective Diels-Alder cycloadditions. Undergoes Friedel-Crafts and cyclocondensation reactions to form bis(pentamethylphenyl)acetic acid and a β-carboline, respectively.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of the American Chemical Society, 109, 3378-3378 (1987)
The Journal of Organic Chemistry, 58, 3231-3231 (1993)
The Journal of Organic Chemistry, 57, 362-362 (1992)
Paris Grant-Preece et al.
Food chemistry, 215, 292-300 (2016-08-21)
Glyoxylic acid is a tartaric acid degradation product formed in model wine solutions containing iron and its production is greatly increased by exposure to UV-visible light. In this study, the combined effect of sulfur dioxide, caffeic acid, pH and temperature
Paris Grant-Preece et al.
Food chemistry, 243, 239-248 (2017-11-18)
Model wine solutions containing organic acids, individually or combined, and iron(III), were exposed to light from fluorescent lamps or stored in darkness for four hours. (-)-Epicatechin was then added, and the solutions incubated in darkness for 10days. Browning was monitored
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service