Skip to Content
Merck
All Photos(2)

Key Documents

E11803

Sigma-Aldrich

2-Ethylaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
1841268
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.2 (vs air)

Quality Level

vapor pressure

0.11 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.559 (lit.)

bp

210 °C (lit.)

mp

−44 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1N

InChI

1S/C8H11N/c1-2-7-5-3-4-6-8(7)9/h3-6H,2,9H2,1H3

InChI key

MLPVBIWIRCKMJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nophawan Paradee et al.
Materials science & engineering. C, Materials for biological applications, 118, 111346-111346 (2020-12-02)
Transdermal drug delivery systems (TDDS) are used as an alternative route to deliver drugs into the blood system for therapy. The matrix materials that have been widely used in TDDS are hydrogels. The dextran hydrogels were prepared by the solution
Taiki Katayama et al.
The ISME journal, 9(2), 436-446 (2014-08-12)
The Minami-Kanto gas field, where gases are dissolved in formation water, is a potential analogue for a marine gas hydrate area because both areas are characterized by the accumulation of microbial methane in marine turbidite sand layers interbedded with mud
Jean Claude Alvarez et al.
Bioanalysis, 7(13), 1675-1684 (2015-08-01)
Remifentanil and propofol are often used in combination for general anesthesia. We developed a method using LC-MS for their simultaneous measurement in human plasma. Methodology & results: After addition of remifentanil-(13)C6 and propofol-d18 (IS), 500 µl of plasma were extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service