DWK92812F-5
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (5 mL), class A
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
PTFE stopper
glass flask
feature
class A
packaging
case of 6 ea
manufacturer/tradename
Kimble® 92812F-5
flask capacity
(5 mL)
General description
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Features and Benefits
- Manufactured from borosilicate glass 3.3
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Heavy duty walls provide superior strength, durability and safety
- Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Printed large volume numbers allow quick identification when selecting glassware
- Flat interior bottoms for stirring with standard stir bars
- Calibrate to contain
- Graduated to Class A volumetric tolerances
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)