Skip to Content
Merck
All Photos(2)

Key Documents

D211605

Sigma-Aldrich

2,2-Diphenylpropionic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H5)2CO2H
CAS Number:
Molecular Weight:
226.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
95%

Quality Level

Assay

95%

form

powder

bp

300 °C (lit.)

mp

172-175 °C (lit.)

SMILES string

CC(C(O)=O)(c1ccccc1)c2ccccc2

InChI

1S/C15H14O2/c1-15(14(16)17,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11H,1H3,(H,16,17)

InChI key

ODELFXJUOVNEFZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R K Gordon et al.
Molecular pharmacology, 36(5), 766-772 (1989-11-01)
Quantitative structure-activity relationships between pharmacological activities and physical properties of a series of 2,2-diphenylpropionate compounds were used to define the topography of the antagonist binding site of muscarinic receptors. XICAMM, a computer molecular modeling program, was used to calculate geometrical
N Toyomura et al.
Bioscience, biotechnology, and biochemistry, 64(3), 610-612 (2000-05-10)
A variety of 2,2-diphenylpropionate derivatives with an amino substituent were synthesized and their effects on larval growth of the silkworm, Bombyx mori, were examined by dietary administration. Of the compounds tested, 3-(4-ethylpiperazin-1-yl)propyl 2,2-diphenylpropionate (3) caused significant prolongation of the larval
T F Holzman et al.
Biochemistry, 20(19), 5524-5528 (1981-09-15)
We have found a new class of inhibitors of the bacterial bioluminescence reaction, the N,N-diphenylalkylamines and acids. We have studied the action of one of these compounds 2,2-diphenylpropylamine. The amine was competitive with the long-chain aliphatic aldehyde substrate (Ki congruent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service