Skip to Content
Merck
All Photos(1)

Key Documents

CF0015

Sigma-Aldrich

4-Bromophenoxytetrafluorobromoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H4Br2F4O
CAS Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

FC(F)(Br)C(F)(F)Oc1ccc(cc1)Br

InChI

1S/C8H4Br2F4O/c9-5-1-3-6(4-2-5)15-8(13,14)7(10,11)12/h1-4H

InChI key

MHPXJSIMKFXETB-UHFFFAOYSA-N

Application

4-Bromophenoxytetrafluoroethyl bromide is a fluoroalkylbromide that is a radical source of the 4-bromophenoxytetrafluoroethyl moiety. Alternatively, it can also be selectively metallated at the fluorinated carbon with Turbo Grignard reagent at low temperatures resulting in a thermally unstable anion that can act as a nucleophilic fluoroalkylation reagent towards a wide variety of electrophiles, such as aldehydes, ketones or sulfonylimines.The aromatic bromide can be further subjected to Pd-catalyzed cross-coupling reactions to build molecular complexity.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service