Skip to Content
Merck
All Photos(1)

Key Documents

C195

Sigma-Aldrich

(2S,3R,4S)-α-(Carboxycyclopropyl)glycine

solid

Synonym(s):

(2S,1′R,2′S)-2-(2-Carboxycyclopropyl)glycine, (2S,3R,4S)-CCG, L-CCG-IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO4
CAS Number:
Molecular Weight:
159.14
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]/D +69.5°, c = 0.4 in H2O(lit.)

reaction suitability

reaction type: solution phase peptide synthesis

color

off-white

solubility

H2O: soluble

application(s)

peptide synthesis

SMILES string

[H][C@]1(C[C@@H]1C(O)=O)[C@H](N)C(O)=O

InChI

1S/C6H9NO4/c7-4(6(10)11)2-1-3(2)5(8)9/h2-4H,1,7H2,(H,8,9)(H,10,11)/t2-,3+,4+/m1/s1

InChI key

GZOVEPYOCJWRFC-UZBSEBFBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent NMDA receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Llansola et al.
Neuroscience, 133(1), 185-191 (2005-05-17)
Hyperammonemia impairs signal transduction associated to glutamate receptors and phosphorylation of some neuronal proteins including microtubule-associated protein 2 (MAP-2). The aim of this work was to analyze the effects of hyperammonemia on modulation of MAP-2 phosphorylation by metabotropic glutamate receptors
Michael Meredith et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(25), 5719-5725 (2004-06-25)
Chemosignals related to reproductive and social status (pheromones) carry messages between opposite-sex and same-sex individuals in many species. Each individual must distinguish signals relevant to its own social behavior with conspecifics from signals used by other (heterospecific) species relevant to
Chao-Yin Chen et al.
The Journal of physiology, 562(Pt 2), 535-551 (2004-11-13)
The nucleus tractus solitarii (NTS) is essential for coordinating arterial baroreflex control of blood pressure. The primary baroreceptor afferent fibres make their first excitatory synaptic contact at second-order NTS neurones with glutamate as the major neurotransmitter. Glutamate regulates its own
Chew L Lau et al.
Neurochemical research, 35(5), 735-742 (2010-02-10)
Astrocytic excitatory amino acid transporters (EAATs) regulate excitatory transmission and limit excitotoxicity. Evidence for a functional interface between EAATs and glial fibrillary acidic protein (GFAP) relevant to astrocytic morphology led to investigations of actions of transportable (D-Aspartate (D-Asp) and (2S,3S,4R)-2-(carboxycyclopropyl)glycine
Joanna M Wierońska et al.
Pharmacological reports : PR, 57(6), 734-743 (2005-12-31)
Several lines of evidence indicate that activation of group II and III metabotropic glutamate (mGlu) receptors produces anxiolytic-like effects in rodents. On the other hand neuropeptide Y (NPY) induces an anxiolytic effect in rats after intraventricular or intraamygdalar administration. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service