Skip to Content
Merck
All Photos(1)

Documents

BR781741

BRAND® microplate BRANDplates®, immunoGrade

size 384 wells, size 100 μL, flat bottom, white

Synonym(s):

BRANDplates®, Immunology, Microplates, high binding, immunoGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

flat bottom
polystyrene
white wells

sterility

non-sterile

feature

lid: no
skirt (F-bottom)

packaging

pack of 50 ea

manufacturer/tradename

BRAND 781741

size

100 μL
384 wells

working volume

100 μL

color

white

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® ImmunoGrade is specifically designed for high-binding capacity, optimized for the immobilization of IgG. This microplate offers superior binding for molecules with mixed hydrophilic and hydrophobic regions, making it the surface of choice for the majority of standard ELISAs and solid-phase immunoassays. Comparable to high-binding plates from other manufacturers, it is made from durable polystyrene (PS) and is non-sterile. Additionally, it is BIO-CERT® CERTIFIED QUALITY, ensuring it is free from human DNA, RNase, DNase, and pyrogens, making it ideal for sensitive applications in research and diagnostics. This microplate is an essential tool for achieving reliable and reproducible results in immunological assays.

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service