Skip to Content
Merck
All Photos(1)

Key Documents

BMSR20200507

Benchmark RotoMini Rotator Tube holder

to hold, 5 x 57 mL

Synonym(s):

tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.11

feature

to hold 5 x 57 mL

manufacturer/tradename

Benchmark Scientific R2020-0507

Looking for similar products? Visit Product Comparison Guide

General description

Additional tube holders to fit the Benchmark RotoMini rotator.

Application

Mixing of samples in tubes

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service