B8908
α-Benzoin oxime
98%
Synonym(s):
alpha-Benzoin oxime, Cuprone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H5CH(OH)C(=NOH)C6H5
CAS Number:
Molecular Weight:
227.26
Beilstein:
1912794
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
153-155 °C (lit.)
storage temp.
2-8°C
SMILES string
O\N=C(\C(O)c1ccccc1)c2ccccc2
InChI
1S/C14H13NO2/c16-14(12-9-5-2-6-10-12)13(15-17)11-7-3-1-4-8-11/h1-10,14,16-17H/b15-13+
InChI key
WAKHLWOJMHVUJC-FYWRMAATSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service