A80602
5-Aminotetrazole monohydrate
97%
Synonym(s):
Tetrazol-5-amine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
CH3N5 · H2O
CAS Number:
Molecular Weight:
103.08
Beilstein:
108872
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
198-204 °C (dec.) (lit.)
SMILES string
[H]O[H].Nc1nnn[nH]1
InChI
1S/CH3N5.H2O/c2-1-3-5-6-4-1;/h(H3,2,3,4,5,6);1H2
InChI key
JVSMPWHQUPKRNV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Useful reagent in condensation reactions to form, for example, pyrimidines.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Indian J. Chem. B, 32, 886-886 (1993)
Annamaria Halasz et al.
Chemosphere, 241, 125008-125008 (2019-10-12)
Tetrazoles have wide industrial applications, notably in the pharmaceutical industry. Tetrazole derivatives such as bis(1H-tetrazol-5-yl)amine (H2BTA) have recently been considered by the defense industry as high nitrogen composite propellants. Photodegradation studies under solar simulating conditions showed that H2BTA was partially
Journal of Heterocyclic Chemistry, 30, 1267-1267 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service