935050
AC1 Screen Printed Electrodes
20 units (2mm Gold Working; Gold Counter; Silver/Silver Chloride Reference)
Synonym(s):
SP-1102
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
The sensor is formed on a corundum ceramic base. On to this surface the working, the reference and the auxiliary electrodes are applied. The working and the auxiliary electrodes are made of variety of materials. At the end of the sensor there is a contacting field which is connected with the active part by the silver conducting paths which are covered by a dielectric protection layer. A biochemically active substance can be immobilized on the working electrode of the sensor to create a biosensor.
We offer Screen Printed Electrodes (2mm Gold Working; Gold Counter; Silver/Silver Chloride Reference) for R&D applications. The physical parameters are as follows:
Dimensions: Weight: 0.5 gm; Length: 25.40 mm; Width: 7.26 mm; Thickness: 0.63 mm
We offer Screen Printed Electrodes (2mm Gold Working; Gold Counter; Silver/Silver Chloride Reference) for R&D applications. The physical parameters are as follows:
Dimensions: Weight: 0.5 gm; Length: 25.40 mm; Width: 7.26 mm; Thickness: 0.63 mm
Application
Screen printed electrodes (2mm Gold Working; Gold Counter; Silver/Silver Chloride Reference) are ideal for electrochemical detection. The various application areas of Screen printed electrodes (SPEs) are:
(1) for the detection of common environmental pollutants such as metal ions and organic compounds;
(2) as enzymatic biosensors, immunosensors and DNA-based sensors for pharmaceutical and clinical usage;
(3) also in emerging applications of fabricating lithium-ion batteries, solar cells, and large area LED matrix.
(1) for the detection of common environmental pollutants such as metal ions and organic compounds;
(2) as enzymatic biosensors, immunosensors and DNA-based sensors for pharmaceutical and clinical usage;
(3) also in emerging applications of fabricating lithium-ion batteries, solar cells, and large area LED matrix.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
High performance screen-printed electrodes prepared by a green solvent approach for lithium-ion batteries
Journal of Power Sources, 334, 65-67 (2016)
Screen printing of stretchable electrodes for large area LED matrix
Journal of Materials Research, 30, 2271-2278 (2015)
Screen printed silver top electrode for efficient inverted organic solar cells
Materials Research Bulletin, 70, 412-415 (2015)
Applications of screen-printed electrodes in current environmental analysis
Current Opinion in Electrochemistry, 3, 137-143 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service