Skip to Content
Merck
All Photos(2)

Key Documents

934747

Sigma-Aldrich

Sulfur

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

Sulfur, Sulfur cathode, cathode for lithium-sulfur batteries

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.21

material

aluminum substrate

Quality Level

grade

battery grade

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

description

2.1 V vs Li/Li+
Description/Nominal Voltage: 2.1 V vs Li/Li+

Assay

≥98%

form

sheet

autoignition temp.

450 °F

composition

Active material loading, 3.75 mg/cm2 ±5% by

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

2E23 μΩ-cm, 20°C

size

5 in. × 10 in.

thickness

16 μm , aluminum current collector
40 μm , excluding current collector
55 μm

capacity

3.0 mAh/cm2 ±5 % (Areal)
(800 mAh/g)

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our sublimed sulfur electrode sheet is a ready-to-use cathode for lithium-sulfur (Li-S) battery research. The sulfur film is cast single-sided on a 16-µm thick carbon-coated aluminum foil current collector that is 5 in. x 10 in. (127 mm x 254 mm) in size. The composition is 70% sublimed sulfur, 10% Poly(vinylidene fluoride) [PVDF] and 20% carbon black.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Our sublimed sulfur electrode sheet is primarily used as a cathode in lithium-sulfur batteries due to its high theoretical capacity. This sulfur cathode can convert sulfur to lithium sulfide by a conversion reaction, resulting in a practical capacity of 800 mAh/g that is 5× than those of the transition-metal oxide cathodes. It offers consistent, reliable performance and is suitable for experimental studies in lithium-sulfur (Li-S) battery chemistry.
battery manufacturing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Sheng Su et al.
Physical chemistry chemical physics : PCCP, 14(42), 14495-14499 (2012-10-04)
Realization of a ubiquitous clean energy future depends critically on the efficient storage and utilization of renewable energies. Lithium-ion batteries are appealing in this regard, but low-cost, abundant, safe, high energy-density electrode materials need to be developed to adopt them.
Cathode Materials for Lithium Sulfur Batteries: Design, Synthesis, and Electrochemical Performance
Duan Lianfeng et al.
Alkali-ion Batteries (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service