Skip to Content
Merck
All Photos(2)

Key Documents

934011

Sigma-Aldrich

Bis(2,2,2-trifluoroethyl) carbonate

greener alternative

≥99%, anhydrous, battery grade

Synonym(s):

BTC, BTFC, Bistrifluoroethyl carbonate, Carbonic Acid Bis(2,2,2-trifluoroethyl) Ester, Carbonic Acid;2,2,2-trifluoroethanol, DFDEC, TFEC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4F6O3
CAS Number:
Molecular Weight:
226.07
EC Number:
807-996-3
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

battery grade

Quality Level

description

Application: Battery manufacturing

Assay

≥99%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm H2O
≤250 ppm HF

bp

118 °C

density

1.51 g/mL

application(s)

battery manufacturing

greener alternative category

SMILES string

FC(F)(F)COC(=O)OCC(F)(F)F

InChI

1S/C5H4F6O3/c6-4(7,8)1-13-3(12)14-2-5(9,10)11/h1-2H2

InChI key

WLLOZRDOFANZMZ-UHFFFAOYSA-N

General description

Bis(2,2,2-trifluoroethyl) carbonate (DFDEC) is a colorless, odorless, and low viscosity liquid widely used in the battery industry as a high-performance electrolyte additive. Its excellent solubility and compatibility with other electrolyte components combined with its high thermal and electrochemical stability ensure outstanding performance in high-temperature and high-voltage applications. Our DFDEC is a highly pure and anhydrous chemical compound manufactured with a minimum purity of 99%.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Bis(2,2,2-trifluoroethyl) carbonate (DFDEC) is a highly effective electrolyte additive that is widely used in the battery industry to enhance the performance and stability of lithium-ion batteries. Its remarkable thermal and electrochemical stability make it an excellent choice for use in high-temperature and high-voltage applications. Recent studies have also highlighted other benefits of DFDEC in battery applications. For instance, it has been found that DFDEC can improve the cycle performance of lithium metal cells by forming a LiF-rich solid electrolyte interphase (SEI).[1,2] Additionally, flammability tests have shown that electrolytes containing over 60% v/v of DFDEC do not catch fire, demonstrating its potential as a flame-retardant.[3] These findings highlight the versatility and importance of DFDEC in the development of safe and high-performance battery technologies.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Non-flammable liquid electrolytes for safe batteries
Gond, R., et al.
Royal Society of Chemistry, 8, 2913-2928 (2021)
Stabilizing the Solid Electrolyte Interphase Layer and Cycling Performance of Silicon-Graphite Battery Anode by Using a Binary Additive of Fluorinated Carbonates
Jo, Hyuntak, et al.
The Journal of Physical Chemistry C, 120, 22466-22475 (2016)
Comparative study of fluorination effect on the linear carbonate molecule for reversible cycling of lithium metal batteries
Lee, et al.
Journal of Electroanalytical Chemistry, 905, 115900-115900 (2022)

Articles

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service