Skip to Content
Merck
All Photos(2)

Key Documents

920460

Sigma-Aldrich

GPhos Pd G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H66NO5PPdS
CAS Number:
Molecular Weight:
906.50
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

O=S(=O)([O-][Pd+2]1([C-]=2C=CC=CC2C=3C=CC=CC3[NH2]1)[P](C=4C(OC(C)(C)C)=CC=C(OC)C4C=5C(=CC=CC5C(C)C)C(C)C)(C6CCCCC6)C7CCCCC7)C

InChI

InChI=1S/C35H53O2P.C12H10N.CH4O3S.Pd/c1-24(2)28-20-15-21-29(25(3)4)32(28)33-30(36-8)22-23-31(37-35(5,6)7)34(33)38(26-16-11-9-12-17-26)27-18-13-10-14-19-27;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h15,20-27H,9-14,16-19H2,1-8H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;-1;;+2/p-1

InChI key

PLMOFCQYSAAMKA-UHFFFAOYSA-M

Application

GPhos Pd G3 is a third generation (G3) Buchwald palladium precatalyst for cross-coupling, demonstrating improved stability and reactivity from previous generations.

Learn more about G3 and G4 Buchwald Precatalysts

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott D McCann et al.
Journal of the American Chemical Society, 142(35), 15027-15037 (2020-08-14)
We have developed a new dialkylbiaryl monophosphine ligand, GPhos, that supports a palladium catalyst capable of promoting carbon-nitrogen cross-coupling reactions between a variety of primary amines and aryl halides; in many cases, these reactions can be carried out at room

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service