Skip to Content
Merck
All Photos(1)

Key Documents

918873

Sigma-Aldrich

Ethanol-d6

reagent grade, ≥98 atom % D, 95% in D2O

Synonym(s):

Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2OD
CAS Number:
Molecular Weight:
52.11
Beilstein:
1699096
MDL number:
UNSPSC Code:
12142201

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

grade

reagent grade

isotopic purity

≥98 atom % D

concentration

95% in D2O

refractive index

n20/D 1.358 (lit.)

bp

78 °C (lit.)

density

0.892 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3,2D2,3D

InChI key

LFQSCWFLJHTTHZ-LIDOUZCJSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service