Skip to Content
Merck
All Photos(1)

Key Documents

913073

Sigma-Aldrich

Hexane-1,6-diammonium iodide

Synonym(s):

1,6-Hexanediamine, dihydriodide (8CI,9CI), Greatcell Solar®, Hexamethylenediamine dihydroiodide, Hexamethylenediammonium diiodide, Hexamethylenediammonium iodide, NHEXDAI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H18I2N2
CAS Number:
Molecular Weight:
372.03
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

form

powder

Quality Level

color

off-white

SMILES string

[I-].[I-].[N+H3]CCCCCC[N+H3]

InChI

1S/C6H16N2.2HI/c7-5-3-1-2-4-6-8;;/h1-8H2;2*1H

InChI key

UBZXDFQTNLTDMC-UHFFFAOYSA-N

Application

Organohalide based perovskites have emerged as an important class of material for solar cell applications. The variations/substitution in organohalide cations and anions is employed for the optimization of the band gap, carrier diffusion length, and power conversion efficiency of perovskites based solar cells.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limeng Ni et al.
ACS nano, 11(11), 10834-10843 (2017-10-25)
Self-assembled hybrid perovskite quantum wells have attracted attention due to their tunable emission properties, ease of fabrication, and device integration. However, the dynamics of excitons in these materials, especially how they couple to phonons, remains an open question. Here, we
Qi Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 30(18), e1704055-e1704055 (2018-03-27)
2D organic-inorganic hybrid perovskites (OIHPs) represent a unique class of materials with a natural quantum-well structure and quasi-2D electronic properties. Here, a versatile direct solution-based synthesis of mono- and few-layer OIHP nanosheets and a systematic study of their electronic structure
Systematic studies on chain lengths, halide species, and well thicknesses for lead halide layered perovskite thin films.
Takeoka Y, et al.
Bulletin of the Chemical Society of Japan, 79(10), 1607-1613 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service