Skip to Content
Merck
All Photos(1)

Key Documents

907073

Sigma-Aldrich

PO-T2T

>=99% (HPLC), Mw 909.8 g/mol,

Synonym(s):

(1,3,5-Triazine-2,4,6-triyl)tris(benzene-3,1-diyl)tris(diphenylphosphine oxide), 2,4,6-Tris[3-(diphenylphosphinyl)phenyl]-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H42N3O3P3
CAS Number:
Molecular Weight:
909.88
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Product Name

PO-T2T, >=99% (HPLC),

grade

sublimed grade (>99% (HPLC))

description

PL-295, 379 nm (in CH2Cl2)

form

solid

mol wt

Mw 909.8 g/mol

color

white

λmax

272 nm in dichloromethane

Orbital energy

HOMO -6.8 eV 
LUMO -2.8 eV 

storage temp.

15-25°C

SMILES string

[P](=O)(c3cc(ccc3)c4nc(nc(n4)c8cc(ccc8)[P](=O)(c%10ccccc%10)c9ccccc9)c5cc(ccc5)[P](=O)(c7ccccc7)c6ccccc6)(c2ccccc2)c1ccccc1

Application

Device configuration:
ITO/MoO3 (3 nm)/ TAPC (35 nm)/CBP:TTM-3NCz (3.0 wt%; (40 nm) and CBP:TTM3PCz (3.0 wt%; 25 nm)/B3PYMPM (10 nm)/PO-T2T (70 nm)/LiF (0.8 nm)/Al (100 nm).

The first Tandem, All-exciplex-based WOLED was constructed using PO-T2T for a device structure configured by two parallel blend layers of mCP/PO-T2T and DTAF/PO-T2T, generating blue and yellow exciplex emission, respectively. The resulting device demonstrates for the first time a tandem, all-exciplex-based white-light OLED (WOLED) with excellent efficiencies ηext: 11.6%, ηc: 27.7 cd/A, and ηp: 15.8 ml/W with CIE(0.29, 0.35) and CRI 70.6 that are nearly independent of EL intensity. The tandem architecture and blend-layer donor/acceptor (1:1) configuration are two key elements that fully utilize the exciplex delay fluorescence, providing a paragon for the use of low-cost, abundant organic compounds en route to commercial WOLEDs.
PO-T2T is an electron transport/hole blocking material in OLED, or an electron acceptor material in organic electronics.

An efficient action of radical-based OLEDs, whose emission originates from a spin doublet, rather than a singlet or triplet exciton was demonstrated in a study published in Nature.
While the emission process is still spin-allowed in these OLEDs, the efficiency limitations imposed by triplet excitons are circumvented for doublets. Using a luminescent radical emitter, an OLED with maximum external quantum efficiency of 27% at a wavelength of 710nm-the highest reported value for deep-red and infrared LEDs was achieved. In this very efficient OLEDs, selective hole injection into the HOMO and electron injection to the SOMO was achieved imploying TAPC as the hole transport layer, and PO-T2T as the electron transport layer to form the fluorescent doublet excited state with near-unity internal quantum efficiency.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Pander et al.
ACS applied materials & interfaces, 10(34), 28796-28802 (2018-07-26)
The photophysics of an exciplex state formed between a small molecule and a polymer is investigated in this work. The results obtained with this blend show the strong potential of polymer-small-molecule blends for triplet harvesting in organic light-emitting diodes (OLEDs)
Yi-Tzu Hung et al.
ACS applied materials & interfaces, 10(40), 34435-34442 (2018-09-18)
Two new nonconjugated linked dicarbazole materials, dCzPSi and dCzPSO2, with high triplet energy were synthesized and characterized. dCzPSi and dCzPSO2 were adopted as unipolar host materials for the green thermally activated delayed fluorescence (TADF) emitter (4CzIPN) to achieve high-efficiency organic
Wen-Yi Hung et al.
Scientific reports, 4, 5161-5161 (2014-06-05)
Exploiting our recently developed bilayer interface methodology, together with a new wide energy-gap, low LUMO acceptor (A) and the designated donor (D) layers, we succeeded in fabricating an exciplex-based organic light-emitting diode (OLED) systematically tuned from blue to red. Further
Tzu-Chieh Lin et al.
Nature communications, 9(1), 3111-3111 (2018-08-08)
The lack of structural information impeded the access of efficient luminescence for the exciplex type thermally activated delayed fluorescence (TADF). We report here the pump-probe Step-Scan Fourier transform infrared spectra of exciplex composed of a carbazole-based electron donor (CN-Cz2) and
Xin Ai et al.
Nature, 563(7732), 536-540 (2018-11-23)
Organic light-emitting diodes (OLEDs)1-5, quantum-dot-based LEDs6-10, perovskite-based LEDs11-13 and micro-LEDs14,15 have been championed to fabricate lightweight and flexible units for next-generation displays and active lighting. Although there are already some high-end commercial products based on OLEDs, costs must decrease whilst maintaining high

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service