Skip to Content
Merck
All Photos(2)

Key Documents

901745

Sigma-Aldrich

5-Bromo-N-(4-fluorobenzyl)-8-hydroxy-1,6-naphthyridine-7-carboxamide

≥90%

Synonym(s):

Aryl halide chemistry informer library compound X10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11BrFN3O2
CAS Number:
Molecular Weight:
376.18
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

Assay

≥90%

form

solid

functional group

amide
bromo
fluoro

SMILES string

Fc1ccc(cc1)CNC(=O)c2nc(c3c(nccc3)c2O)Br

InChI key

VRVGOUHPADQNHC-UHFFFAOYSA-N

Application

This product is Informer compound X10 of the Aryl Halide Chemistry Informer Library developed by chemists at Merck & Co., Inc., Kenilworth, NJ, U.S., which contains 18 drug-like molecules representative of those encountered in complex synthesis. By screening a new reaction against the Informer Library, chemists can directly compare and analyse a reaction′s successes and shortcomings among different methods and various research teams. It may also be used to facilitate deeper method development for performance or utility.

Caution

Not Fully Tested.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter S Kutchukian et al.
Chemical science, 7(4), 2604-2613 (2016-04-21)
Major new advances in synthetic chemistry methods are typically reported using simple, non-standardized reaction substrates, and reaction failures are rarely documented. This makes the evaluation and choice of a synthetic method difficult. We report a standardized complex molecule diagnostic approach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service