Skip to Content
Merck
All Photos(2)

Key Documents

799106

Sigma-Aldrich

2-Hydroxypropyl methacrylamide

99% (GC)

Synonym(s):

N-(2-Hydroxypropyl)-2-methyl-prop-2-enamide, N-(2-hydroxypropyl)methacrylamide), HPMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99% (GC)

form

powder

mp

67.92 °C

density

1.002 g/mL (predicted)

storage temp.

2-8°C

SMILES string

CC(O)CNC(=O)C(C)=C

InChI

1S/C7H13NO2/c1-5(2)7(10)8-4-6(3)9/h6,9H,1,4H2,2-3H3,(H,8,10)

InChI key

OKPYIWASQZGASP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monomers to synthesize biocompetible polymers and copolymers for drug carriers, protein conjugates and hydrogels in biomedical applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jindrich Kopecek et al.
Advanced drug delivery reviews, 62(2), 122-149 (2009-11-19)
The overview covers the discovery of N-(2-hydroxypropyl)methacrylamide (HPMA) copolymers, initial studies on their synthesis, evaluation of biological properties, and explorations of their potential as carriers of biologically active compounds in general and anticancer drugs in particular. The focus is on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service