Skip to Content
Merck
All Photos(1)

Documents

768529

Sigma-Aldrich

Lithium-6Li oxide

95 atom % 6Li, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
6Li2O
Molecular Weight:
28.03
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

isotopic purity

95 atom % 6Li

Assay

97%

form

powder

mass shift

M+1

SMILES string

[6Li+].[6Li+].[O-2]

InChI

1S/2Li.O/q2*+1;-2/i2*1-1;

InChI key

FUJCRWPEOMXPAD-NQTTVQCTSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service