Skip to Content
Merck
All Photos(1)

Key Documents

760668

Sigma-Aldrich

Dibenzocyclooctyne-maleimide

for Copper-free Click Chemistry

Synonym(s):

DBCO-maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O4
Molecular Weight:
427.45
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

maleimide

storage temp.

−20°C

SMILES string

O=C(N1CCC(NCCC(N2CC3=C(C=CC=C3)C#CC4=C2C=CC=C4)=O)=O)C=CC1=O

InChI

1S/C25H21N3O4/c29-22(14-16-27-23(30)11-12-24(27)31)26-15-13-25(32)28-17-20-7-2-1-5-18(20)9-10-19-6-3-4-8-21(19)28/h1-8,11-12H,13-17H2,(H,26,29)

InChI key

NHFQNAGPXIVKND-UHFFFAOYSA-N

Application

Maleimide functionalized cyclooctyne derivative for incorporation of the cyclooctyne moiety into thiol containing compounds or biomolecules. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage.

Applications Include:
  • Protein-peptide conjugates
  • Antibody-enzyme or antibody-drug conjugates
  • Protein or peptide-oligonucleotide conjugates
  • Surface modification

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative study of bioorthogonal reactions with azides
Agard, N.; et al.
ACS Chemical Biology, 10, 644-648 (2006)
Highly robust and optimized conjugation of antibodies to nanoparticles using quantitatively validated protocols.
Jeong S, et al.
Nanoscale, 9(7), 2548-2555 (2017)
Site-selective installation of BASHY fluorescent dyes to Annexin V for targeted detection of apoptotic cells
Cal PM, et al.
Chemical Communications (Cambridge, England), 53(2), 368-371 (2017)
Hansjörg Götzke et al.
Nature communications, 10(1), 4403-4403 (2019-09-29)
Specialized epitope tags are widely used for detecting, manipulating or purifying proteins, but often their versatility is limited. Here, we introduce the ALFA-tag, a rationally designed epitope tag that serves a remarkably broad spectrum of applications in life sciences while
Sebastian Strauss et al.
Nature methods, 17(8), 789-791 (2020-07-01)
DNA-PAINT's imaging speed has recently been significantly enhanced by optimized sequence design and buffer conditions. However, this implementation has not reached an ultimate speed limit and is only applicable to imaging of single targets. To further improve acquisition speed, we

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service