752185
Benzyl diazoacetate solution
10% in toluene
Synonym(s):
Benzyl 2-diazoacetate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H8N2O2
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form
liquid
concentration
10% in toluene
refractive index
n20/D 1.504
density
0.894 g/mL at 25 °C
functional group
ester
phenyl
storage temp.
2-8°C
SMILES string
[N-]=[N+]=CC(=O)OCc1ccccc1
InChI
1S/C9H8N2O2/c10-11-6-9(12)13-7-8-4-2-1-3-5-8/h1-6H,7H2
InChI key
JBBKWIFIXPBQGZ-UHFFFAOYSA-N
Related Categories
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
45.0 °F
Flash Point(C)
7.2 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service