Skip to Content
Merck
All Photos(2)

Key Documents

745480

Sigma-Aldrich

Zinc isopropylsulfinate

95%

Synonym(s):

Baran Isopropylation Reagent, Baran IPS Reagent, Bis((isopropylsulfinyl)oxy)zinc, IPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O4S2Zn
CAS Number:
Molecular Weight:
279.69
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: Allylation
reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

reagent type: diversification reagent

mp

161-166 °C

functional group

sulfinic acid

SMILES string

CC(C)S(=O)O[Zn]OS(=O)C(C)C

InChI

1S/2C3H8O2S.Zn/c2*1-3(2)6(4)5;/h2*3H,1-2H3,(H,4,5);/q;;+2/p-2

InChI key

PEEMMWYYSHSBTQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc isopropylsulfinate (IPS) is part of a zinc sulfinate toolbox developed by Phil Baran for the simple and rapid diversification of heterocycles.[1]


Practical and Innate Carbon-Hydrogen Functionalization of Heterocycles

Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Linkage

Frequently Asked Questions are available for this Product.

Preparation Note

may contain up to 1 mole eq ZnCl2

Other Notes

Previously sold under product number L511161

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rie Poulsen et al.
Trials, 18(1), 578-578 (2017-12-05)
Depression and anxiety are among the largest contributors to the global burden of disease and have negative effects on both the individual and society. Depression and anxiety are very likely to influence the individual's work ability, and up to 40%
Chun You et al.
Biotechnology and bioengineering, 114(8), 1855-1864 (2017-04-15)
Myo-Inositol (vitamin B8) is widely used in the drug, cosmetic, and food & feed industries. Here, we present an in vitro non-fermentative enzymatic pathway that converts starch to inositol in one vessel. This in vitro pathway is comprised of four
Elisa Zamboni et al.
eLife, 9 (2020-11-11)
Adapting to the environment statistics by reducing brain responses to repetitive sensory information is key for efficient information processing. Yet, the fine-scale computations that support this adaptive processing in the human brain remain largely unknown. Here, we capitalise on the
Elisabeth Maurer et al.
Medical mycology, 57(3), 351-362 (2018-06-21)
Mucorales can cause cutaneous to deep-seated infections, mainly in the immunocompromised host, resulting in high mortality rates due to late and inefficient treatment. In this study, Galleria mellonella larvae were evaluated as a heterologous invertebrate host to study pathogenicity of
Christopher A Barwacz et al.
The International journal of oral & maxillofacial implants, 33(5), 1126-1135 (2018-09-20)
To evaluate the influence that three different implant-abutment interface designs had on peri-implant mucosal outcomes as assessed by the pink esthetic score (PES) 3 years after delayed implant placement and immediate provisionalization. Adult subjects (n = 141) requiring replacement of

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service